Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ Adhere Columns

Catalog No. p-3676463
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.30 in.
0.62 in.
Flow Rate:
<20 mL/min.
<4 mL/min.
Max. Pressure:
1.5 bar
44 bar

Designed to remove contaminants in post-protein A purification

  • Prepacked with Capto adhere media, a strong anion exchanger with multimodal functionality
  • High capacity and productivity in flow-through mode: 0.09 to 0.12mmol Cl-/mL medium ionic capacity
  • Wide operational window of pH and conductivity: working pH range 3 to 12
  • Two-step process reduces time spent and operating costs

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto adhere column
Bed Volume 1mL
Diameter (English) Inner 0.30 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Ligand Type N-benzyl-n-methyl Ethanolamine
Max. Pressure 1.5 bar
pH Range 3 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Easy screening and convenient process development
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.