Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ PlasmidSelect

Catalog No. 09928117 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume (Metric)
09-928-117 1 mL
09-928-118 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-117 Supplier Cytiva Supplier No. 17549911
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

For the purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications

  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap™ columns
  • Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Purification of Supercoiled DNA From Research Scale to cGMP Production
Product Type Chromatography Resin
Flow Rate <4 mL/min (Max)
Max. Pressure 5 bar (0.5 MPa)
Matrix Capto™ PlasmidSelect
Bed Volume (Metric) 1 mL
Packaging Quantity 5/Pk.
Diameter (Metric) 0.7 cm
Length (Metric) 2.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.