Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Con A 4B Columns

Catalog No. 45002925 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Flow Rate:
<20 mL/min.
<4 mL/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Flow Rate
45-002-925 5 x 1mL 0.27 in. <4 mL/min.
45-002-926 5 x 5mL 0.62 in. <20 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-925 Supplier Cytiva Supplier No. 28952085
Add to Cart
Add to Cart

For separation and purification of glycoproteins, polysaccharides and glycolipids

  • Prepacked with Con A Sepharose™ 4B media
  • Bed volume: 1mL and 5mL
  • Bed dimensions: 7 x 25mm (1mL), 16 x 25mm (5mL)
  • Column material: Polypropylene (PP)
  • Max flow rate: 4mL/min (1mL), 20mL/min (5mL)
Media
  • Ligand: Concanavalin A (Con A)
  • Matrix: 4% agarose
  • Average particle size: 90μm
  • pH stability working range: 4 to 9

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Con A Sepharose 4B column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Ligand Type Concanavalin A (13mg Con A/mL drained medium)
Max. Pressure 3 bar
pH Range 4 to 9
Particle Size 45 to 165μm
Support Type 4% Agarose
Content And Storage 4° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Convenient separation and purification of Glycoproteins, Polysaccharides and Glycolipids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.