Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ HiTrap™ Histidine Tagged Package

Catalog No. 01257020 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-257-020 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-020 Supplier Cytiva™ Supplier No. 29058803
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The His package enables convenient purification of histidine-tagged proteins.

  • Contains HisTrap™ HP (1 x 1 mL) HiTrap™ TALON crude (1 x 1 mL) HiTrap™ Desalting (1 x 5 mL)
  • Comes with ready-to-use columns packed with high performance resins developed for high yield (HisTrap™ HP) or high purity (HiTrap™ TALON™crude)
  • Includes HiTrap™ Desalting column for removing imidazole from eluate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material His Tag Antibody Plate
Product Line HiTrap
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.