Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ IMAC Columns

Catalog No. 45000164 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Particle Size:
34 μm
90 μm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Particle Size
45-000-164 5 x 5 mL 0.62 in. 34 μm
45-000-163 5 x 1 mL 0.27 in. 34 μm
45-000-167 5 x 1 mL 0.27 in. 90 μm
45-000-168 5 x 5 mL 0.62 in. 90 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-000-164 Supplier Cytiva Supplier No. 17092005
Add to Cart
Add to Cart

Includes

Instructions, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems

For optimizing purification of histidine-tagged proteins when Ni +2 is not the best choice of metal ion

  • Conveniently charge with your metal of choice
  • Can be used with a syringe, peristaltic pump, or liquid chromatography system
  • Available prepacked with IMAC Sepharose™ High Performance (HP), with small bead size for high performance purification, or IMAC Sepharose 6 Fast Flow (FF), with larger bead size for easy scaleup
  • High binding capacity: ≥40mg histidine-taged protein/mL medium (protein/metal ion dependent)
  • Stable in the presence of all commonly used buffers
  • Available in 1mL and 5mL sizes

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.62 in.
Flow Rate <4 mL/min.
Diameter (Metric) Inner 16 mm
Kit Contents Instructions, plus a variety of connecto rs for syringe, peristaltic pump and chromato graphy systems
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system
Material Polypropylene
Max. Pressure 42 psi
pH Range 3 to 12
Particle Size 34 μm
Product Type HiTrap IMAC HP Column
Content And Storage 4°C to 30°C
Quantity 5 x 5 mL
For Use With (Application) For optimizing purification of His-tagged Proteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.