Learn More
Description
Includes
Instructions, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems
- Conveniently charge with your metal of choice
- Can be used with a syringe, peristaltic pump, or liquid chromatography system
- Available prepacked with IMAC Sepharose™ High Performance (HP), with small bead size for high performance purification, or IMAC Sepharose 6 Fast Flow (FF), with larger bead size for easy scaleup
- High binding capacity: ≥40mg histidine-taged protein/mL medium (protein/metal ion dependent)
- Stable in the presence of all commonly used buffers
- Available in 1mL and 5mL sizes
Specifications
Specifications
| Bed Volume | 1 mL |
| Diameter (English) Inner | 0.27 in. |
| Flow Rate | <4 mL/min. |
| Diameter (Metric) Inner | 7 mm |
| Kit Contents | Prepacked with IMAC Sepharose 6 Fast flow media |
| For Use With (Equipment) | Syringe, peristaltic pump or liquid chromato graphy system |
| Material | Polypropylene |
| Max. Pressure | 42 psi |
| pH Range | 3 to 12 |
| Particle Size | 90 μm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.