Learn More
Description
- Prepacked with Protein A Sepharose High Performance media for purification and fractionation of IgG subclasses
- For use with syringe, pump or chromatography system such as KTA design or FPLC System
- Binds 20mg human IgG/mL medium
- Stable over pH 3 to 9 for use and 2 to 10 for cleaning
- Designed for use with a syringe, a peristaltic pump, or a chromatography system
- Maximum pressure: 3 bar
- Column material: polypropylene
- 1 and 5mL columns
Media
- Ligand: Native protein A
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: 3 to 9
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Binding Capacity | 20 mg/mL IgG Medium |
| Flow Rate | <4 mL/min. |
| For Use With (Application) | Chromatography System, Manual |
| For Use With (Equipment) | For syringe, peristaltic pump or a chromatography system |
| Content And Storage | 4°C to 8°C, 20% Ethanol |
| Quantity | 2 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.