Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ rProtein A FF Columns

Catalog No. 45000263 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
2 x 1 mL
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate
45-000-263 2 x 1 mL <4 mL/min.
45-000-262 5 x 1 mL <4 mL/min.
45-000-265 5 x 5 mL <20 mL/min.
45-000-264 1 x 5 mL <20 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-000-263 Supplier Cytiva Supplier No. 17507902
Add to Cart
Add to Cart

Use for preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, serum and ascites. Cytiva Life Sciences™ HiTrap™ rProtein A FF Columns are prepacked with rProtein A Sepharose™ Fast Flow.

  • Packed with rProtein A Sepharose™ Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources
  • Exhibits similar Fc region specificity to that of native protein A but with enhanced binding capacity and fewer regulatory concerns due to total absence of mammalian culture in ligand production and purification
  • Average particle size: 90μm
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
  • Available as 1mL and 5mL columns
Media
  • Ligand: Recombinant protein A (E.coli)
  • Matrix: Highly cross-linked 4% agarose
  • Binding capacity: ∽50mg human IgG/mL
  • pH stability working range: 3 to 10

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Order Info

1mL and 5mL columns are packed with 1mL and 5mL of rProtein A Sepharose Fast Flow, respectively.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity ≈50 mg Human IgG/mL
Flow Rate <4 mL/min.
For Use With (Application) Chromatography System, Manual
For Use With (Equipment) For syringe, peristaltic pump or a chromatography system
Content And Storage 4°C to 30°C, 20% Ethanol
Quantity 2 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.