Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ TALON™ Crude Prepacked Columns
GSA_VA

Catalog No. 45002386 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
5 x 1 mL
5 x 5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-386 5 x 5 mL
45-002-385 5 x 1 mL
45-000-502 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-386 Supplier Cytiva Supplier No. 28953767
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC). Cytiva Lifescience™ HiTrap™ TALON™ Crude Prepacked Columns offer a cobalt-based medium allowing protein purification under native or denaturing conditions.

Cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. Allows protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.

  • Prepacked with cobalt-based chromatography media
  • Binding capacity approximately 7mg MBP
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL columns

Media
  • Ligand: TALON™ Superflow Cobalt-IMAC
  • Matrix: 6% cross-linked agarose
  • Average particle size: 60 to 160μm
  • pH stability working range: 3 to 12

1mL columns

  • Column I.D.: 7mm
  • Flow velocity: 1 to 4mL/min.

5mL columns
  • Column I.D.: 16
  • Flow velocity: 5 to 20mL/min.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 5 mL
For Use With (Application) For PurifiCatIon of Histidine-Tagged Recombinant Proteins by Immobilized Metal Affinity Chromatography, Used Buffers and Easily Regenerated with 0.5 M NaOH
Storage Requirements 4°C to 8°C
Binding Capacity Up to 20 mg Histidine-tagged Protein1
Diameter (English) Inner 0.62 in.
Product Type HiTrap Talon crude prepacked Column
Diameter (Metric) Inner 1.6 cm
Flow Rate 1 to 4 mL/min
Particle Size 60 to 160 μm
Material Polypropylene
Max. Pressure 3 bar
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Talon Superflow Cobalt-IMAC
Packaging Quantity 5 x 5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.