Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody (HIV1-24/661), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen HIV-1 p24
Clone HIV1-24/661
Dilution Immunocytochemistry/Immunofluorescence 0.5-1ug/ml
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP234335F
SDP
View Documents
Novus Biologicals
NBP2343350.02MG
0.02 mg
Each for $268.00
Only null left
Add to Cart
 
NBP234335A
SDP
View Documents
Novus Biologicals
NBP2343350.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP234335B
SDP
View Documents
Novus Biologicals
NBP2343350.2MG
0.2 mg N/A N/A N/A
Description

Description

HIV-1 p24 Monoclonal specifically detects HIV-1 p24 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

HIV-1 p24
Immunocytochemistry/Immunofluorescence 0.5-1ug/ml
Monoclonal
Purified
RUO
PBS with 0.05% BSA. with 0.05% Sodium Azide
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24
Recombinant HIV-1 Gag p24 protein
Primary
Store at 4C.
HIV1-24/661
Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Virus
P04585
155348
IgG1 κ
Protein A purified
Human immunodeficiency virus (HIV) is a retrovirus that causes acquired immune deficiency syndrome (AIDS), a condition in humans in which the immune system begins to fail, leading to life-threatening opportunistic infections. HIV mainly infects vital cells in the human immune system such as helper T cells (specifically CD4+ T cells), macrophages and dendritic cells. Two species of HIV infect humans: HIV-1 and HIV-2, with HIV-1 being the more virulent strain. The gag gene of human immunodeficiency virus 1 (HIV-1) encodes a precursor protein known as Pr55Gag. The viral protease PR cleaves this precursor to generate p17, p24, p7, and p6 proteins, which are required for virus particle assembly. HIV-1 Gag p24 is a capsid protein that constitutes the core of AIDS virus HIV-1. p6 and p7 are the components of the nucleocapsid, and p17 provides a protective matrix. HIV-1 Gag p24 is indispensable to the reproduction of AIDS virus and constitutes an essential element for the AIDS virus particle construction. As this protein is detectable from the early stage of AIDS virus infection, its measurement is commonly used as an indicator of HIV-1 infection and viral load.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.