Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Gag p24 Antibody (ND1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500473AF750
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
HIV-1 Gag p24 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
gag | |
HIV-1 Gag p24 C-terminal peptide. | |
Primary | |
The antibody reacts with HIV-1 Gag p24). Specificity was verified by Western Blotting in reducing conditions with Sanofi Pasteur NEW LAV-BLOT I kit. Epitope was reported to be located within the amino acids 51-110. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
ND1 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
Mouse | |
0.1 mL | |
155030 | |
Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction