Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Gag p24 Antibody (ND1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500473V
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.Specifications
HIV-1 Gag p24 | |
Monoclonal | |
DyLight 405 | |
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
Mouse | |
Affinity Purified | |
RUO | |
155030 | |
Virus | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
ND1 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
gag | |
HIV-1 Gag p24 C-terminal peptide. | |
0.1 mL | |
Primary | |
The antibody reacts with HIV-1 Gag p24). Specificity was verified by Western Blotting in reducing conditions with Sanofi Pasteur NEW LAV-BLOT I kit. Epitope was reported to be located within the amino acids 51-110. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction