Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Gag p24 Antibody (ND1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500473H
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
HIV-1 Gag p24 | |
Monoclonal | |
HRP | |
PBS | |
HIV-1 Gag p24 C-terminal peptide. | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
155030 | |
Store at 4C in the dark. |
Western Blot, ELISA, Immunocytochemistry | |
ND1 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
Mouse | |
Multi-step | |
RUO | |
Primary | |
Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction