Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p24 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7360SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7360SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7360SP Supplier R&D Systems Supplier No. MAB7360SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIV-1 Gag p24 Monoclonal specifically detects HIV-1 Gag p24 in Viral samples. It is validated for Western Blot.

Specifications

Antigen HIV-1 Gag p24
Applications Western Blot
Classification Monoclonal
Clone 749140
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_057850
Gene Symbols gag
Host Species Mouse
Immunogen E. coli-derived recombinant viral HIV-1 Gag p24 Pro149-Gly354 Accession # NP_057850
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155030
Test Specificity Detects viral HIV-1 Gag p24 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Virus
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.