Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp120 Antibody (4F4E4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306629G
Description
HIV-1 gp120 Monoclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISASpecifications
HIV-1 gp120 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA | |
4F4E4 | |
Western Blot, ELISA | |
Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 gp120 (Ala 29-Glu 503; 97CN001 strain). | |
0.1 mL | |
Virology Bacteria and Parasites | |
155971 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction