Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP241090
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.Specifications
HIV-1 gp120 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
env | |
Antibody was raised against 15 amino acids from near the center of gp120. The immunogen is located within amino acids 470 - 520 of gp120. Amino Acid Squence: APTKAKRRVVQREKR | |
0.1 mg | |
Primary | |
Virus | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1 ug/ml, ELISA | |
Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
Goat | |
Affinity Purified | |
RUO | |
155971 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction