Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp120 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP241090MFV610
Description
HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISASpecifications
| HIV-1 gp120 | |
| Polyclonal | |
| Western Blot, ELISA | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| Antibody was raised against 15 amino acids from near the center of gp120. The immunogen is located within amino acids 470 - 520 of gp120. Amino Acid Squence: APTKAKRRVVQREKR | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 155971 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Goat | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction