Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp120 (PNDmn) Monoclonal Antibody (ANT-151), QED Bioscience™
SDP

Catalog No. 502278139
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8139 1 mg
50-227-8140 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-227-8139 Supplier Leinco Technologies Inc Supplier No. 560121000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIV-1 gp120 (PNDmn) Monoclonal antibody specifically detects HIV-1 gp120 (PNDmn) in Virus samples. It is validated for ELISA

Specifications

Antigen HIV-1 gp120 (PNDmn)
Applications ELISA
Classification Monoclonal
Clone ANT-151
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias HIV Glycoprotein 120; HIV-1 Glycoprotein 120; Human immunodeficiency virus
Host Species Mouse
Immunogen Recombinant gp120 (MN strain), a CCR5 and X4 tropic strain.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.