Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 gp24 Monoclonal Antibody (ANT-152), QED Bioscience™
SDP

Catalog No. 502278143
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8143 1 mg
50-227-8144 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-227-8143 Supplier Leinco Technologies Inc Supplier No. 560161000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIV-1 gp24 Monoclonal antibody specifically detects HIV-1 gp24 in Virus samples. It is validated for Functional Assay assay, Immunoprecipitation

Specifications

Antigen HIV-1 gp24
Applications Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone ANT-152
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen r.gp24
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 2-8°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.