Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 gp41 Antibody (417D3.08/ clone 2), Alexa Fluor™ 546, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX1305A546
Description
HIV-1 gp41 Monoclonal specifically detects HIV-1 gp41 in Virus samples. It is validated for Flow Cytometry, ELISA.Specifications
HIV-1 gp41 | |
Monoclonal | |
Alexa Fluor 546 | |
Env, Glycoprotein 41, gp41, Human immunodeficiency virus 1, Human Immunodeficiency Virus Type 1 gp41, TM, Transmembrane protein | |
HR1-PID-HR2-transfected HEK 293 cells (HIV-1, 92BR025, Clade C) | |
0.1 mg | |
Primary | |
Store at -20C in the dark. Avoid freeze-thaw cycles. |
Flow Cytometry, ELISA | |
417D3.08/ clone 2 | |
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Human, Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction