Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Rev binding protein (HRB) Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | HIV-1 Rev binding protein (HRB) |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HIV-1 Rev binding protein (HRB) Polyclonal specifically detects HIV-1 Rev binding protein (HRB) in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HIV-1 Rev binding protein (HRB) | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3267 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
ArfGAP with FG repeats 1, DKFZp686I15205, HIV-1 Rev-binding protein, HRBRev interacting protein, Rab, Rev/Rex activation domain-binding protein, RABMGC116938, RIPMGC116940 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HIV-1 Rev binding protein (HRB) (NP_004495.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title