Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 tat Antibody - Azide and BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB12014029
Description
HIV-1 tat Polyclonal specifically detects HIV-1 tat in Non-species specific samples. It is validated for Western Blot.Specifications
HIV-1 tat | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
Human |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:500-1:2000 | |
Tat | |
Full length HIV-1 tat protein (Human). | |
0.05 mg | |
Virology Bacteria and Parasites | |
This may cross-react to various subtypes of HIV-1 tat protein | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction