Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV TAT specific factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30330620UL
Description
HIV TAT specific factor 1 Polyclonal specifically detects HIV TAT specific factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HIV TAT specific factor 1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
27336 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human HIV TAT specific factor 1 (NP_055315.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction