Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix HIV Type 1 p24 Extended Range Kit
Shop All ZeptoMetrix Corporation Products

Click to view available options
Quantity:
Sufficient for 5 x 96 determinations
Description
- Allows samples containing p24 antigen up to 4ng/mL to be accurately quantified without extensive serial dilution
Specifications
Specifications
Content And Storage | 2°C to 8°C |
Includes | Concentrated HIV-1 p24 Antigen Standard (0.3mL), SIGMAFAST™ OPD tablet sets (5 sets) |
Quantity | Sufficient for 5 x 96 determinations |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction