Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV1 Gag Protein Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986251
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6251 Supplier Bioss Supplier No. BS4981R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Matrix protein p17 targets Gag and Gag-Pol polyproteinss to the plasma membrane via a multipartite membrane binding signal. that includes its myristoylated N-terminus. Also mediates nuclear localization of the preintegration complex. Implicated in the release from host cell mediated by Vpu.

Specifications

Antigen HIV1 Gag Protein
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from HIV1 Nucleocapsid protein p7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.