Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 gp41 Monoclonal Antibody (1D4)

Catalog No. 01670324
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-324 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-324 Supplier Invitrogen™ Supplier No. MA17590
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-7590 detects HIV from virus samples. The MA1-7590 immunogen is a recombinant HIV1 gp41 protein. Store at 4C for 3 months or less. For long term store at -80C.

Gp41 is a glycoprotein, that along with gp120 is integral to the process by which HIV enters the cell. Gp41 is primarily located within the viral envelope, however when gp120 binds to a CD4 receptor, gp120 changes its conformation, causing gp41 to become.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 gp41
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias HIV Glycoprotein 41
Host Species Mouse
Immunogen Recombinant HIV1 gp41 protein.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.