Learn More
Invitrogen™ HIV1 gp41 Polyclonal Antibody, Biotin

Description
The PA1-73100 antibody reacts with HIV-1 gp41. PA1-73100 has been successfully used in Western blot applications. The PA1-73100 antibody was raised against Recombinant ectodomain of gp41-glycosylated (a.a. residues 546-682) from strain HxB2. Recommended short term of less than 6 months at 2-8°C. For longer storage, aliquot and freeze.
Specifications
Specifications
| Antigen | HIV1 gp41 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | HIV Glycoprotein 41 |
| Host Species | Goat |
| Immunogen | Recombinant ectodomain of gp41-glycosylated (a.a. residues 546-682) from strain HxB2. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.