Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 gp41 Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA173099
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173099 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173099 Supplier Invitrogen™ Supplier No. PA173099
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73099 antibody reacts with HIV-1 gp41. PA1-73099 has been successfully used in ELISA and WB applications. The PA1-73099 antibody was raised against Recombinant ectodomain of gp41-glycosylated (a.a. residues 546-682) from strain HxB2.

Gp41 is a glycoprotein, that along with gp120 is integral to the process by which HIV enters the cell. Gp41 is primarily located within the viral envelope, however when gp120 binds to a CD4 receptor, gp120 changes its conformation, causing gp41 to become.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 gp41
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1-2 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Gene Alias HIV Glycoprotein 41
Host Species Goat
Immunogen Recombinant ectodomain of gp41-glycosylated (a.a. residues 546-682) from strain HxB2.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.