Learn More
Invitrogen™ HIV1 Virions Polyclonal Antibody

Description
PA1-7216 detects the HIV-1 (HTLV-3). PA1-7216 has been successfully used in immunofluorescence procedures. The PA1-7216 immunogen is human isolate, confirmed.
Specifications
Specifications
| Antigen | HIV1 Virions |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | HIV-1; HIV-1 Virions |
| Host Species | Goat |
| Immunogen | Human isolate, confirmed. |
| Purification Method | IgG Fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.