Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 Integrase Monoclonal Antibody (IN-2)
GREENER_CHOICE

Catalog No. PIMA546563
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546563 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546563 Supplier Invitrogen™ Supplier No. MA546563
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIV1 Integrase Monoclonal Antibody for Western Blot, ICC/IF, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 Integrase
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone IN-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias HIV-1
Host Species Mouse
Immunogen Bacterially expressed, hexahistidine amino-terminal tagged HIV-1 integrase (IN) protein (clade B, HXB-3 isolate).
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.