Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 nef Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14963 Supplier Invitrogen™ Supplier No. PA14963
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

PA1-4963 detects the HIV-1 Nef protein in HIV samples. PA1-4963 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ~27 kDa protein, corresponding to HIV-1 Nef in HIV samples. PA1-4963 immunogen corresponds to the recombinant protein which was over-expressed in E. coli with a plasmid carrying the Nef coding region of the HIV-1 virus. Recombinant HIV-1 Nef protein (Cat. # RP-4922 or RP-4923) can be purchased for control experiments in Western blot assays.

HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD 4 and MH4, important for AIDS virus infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 nef
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.09% sodium azide
Gene Alias HIV-1 Nef
Host Species Guinea Pig
Immunogen Purified full-size recombinant Nef of HIV1 group M, subtype B expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.