Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 p17 Monoclonal Antibody (G35H)
GREENER_CHOICE

Catalog No. MA17041
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17041 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17041 Supplier Invitrogen™ Supplier No. MA17041
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7041 detects HIV-1. MA1-7041 has been successfully used in ELISA and Western blot procedures. The MA1-7014 immunogen is purified recombinant HIV-1 p17.

HIV-1 Gag p17 is the matrix protein of AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitutes an essential element for the of AIDS virus particle construction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 p17
Applications ELISA, Western Blot
Classification Monoclonal
Clone G35H
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HIV1gp1
Host Species Mouse
Immunogen Purified recombinant HIV-1 p17.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.