Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 p17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14954 Supplier Invitrogen™ Supplier No. PA14954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4954 detects the HIV-1 p17 protein in HIV samples. PA1-4954 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ~17 kDa protein, and a ~55 kDa protein, corresponding respectively to HIV-p17 and to its precursor p55, in HIV samples. PA1-4954 immunogen corresponds to the recombinant protein which was over-expressed in E. coli with a plasmid carrying the Gag p17 coding region of the HIV-1 virus. Recombinant HIV-1 Gag p17 protein (Cat. # RP-4914 or RP-4915) can be purchased for control experiments in Western blot assays.

HIV-1 Gag p17 is the matrix protein of AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitutes an essential element for the of AIDS virus particle construction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 p17
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.09% sodium azide
Gene Alias HIV1gp1
Host Species Rabbit
Immunogen Purified full-size recombinant Gag p17 of HIV1 subtype B expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.