Learn More
Invitrogen™ HIV1 p24 Monoclonal Antibody (2N34)

Description
This antibody reacts with HIV-1 p24 subtype B, C and A. It has been mapped to amino acids 74-84 (NEEAAEWDRVH - B subtype consensus). This clone may also be referred to as 2N34 (05-008).
Specifications
Specifications
| Antigen | HIV1 p24 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2N34 |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | HIV p24; HIV1gp1 |
| Host Species | Mouse |
| Immunogen | Recombinant p24 protein of HIV-1 B-subtype consensus purified fromE.coli. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.