Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIV1 p24 Monoclonal Antibody (D96A)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17381
Description
The MA1-7381 antibody reacts with HIV-1 p24 in viral and infected tissue samples. MA1-7381 has been successfully used in ELISA applications. The MA1-7381 antibody was raised against HIV protein 24.
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Specifications
HIV1 p24 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA | |
D96A | |
Unconjugated | |
HIV p24; HIV1gp1 | |
HIV protein 24. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction