Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 p24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581773 Supplier Invitrogen™ Supplier No. PA581773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for HIV-1 Gag-P24.

HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 p24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias HIV p24; HIV1gp1
Host Species Rabbit
Immunogen Recombinant HIV-1 Gag-P24 protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.