Learn More
Invitrogen™ HIV1 p24 Polyclonal Antibody, HRP

Description
The PA1-73094 antibody reacts with HIV-1 p24. PA1-73094 has been successfully used in ELISA applications. The PA1-73094 antibody was raised against purified native p24 from strain IIIB. NOTE: sodium azide is a potent inhibitor of peroxidase.
Specifications
Specifications
| Antigen | HIV1 p24 |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal |
| Gene Alias | HIV p24; HIV1gp1 |
| Host Species | Goat |
| Immunogen | Purified native p24 from strain IIIB. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.