Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 Reverse Transcriptase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA14948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA14948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA14948 Supplier Invitrogen™ Supplier No. PA14948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4948 detects the HIV-1 Reverse Transcriptase protein in HIV samples. PA1-4948 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ~66 kDa protein and a ~51 kDa protein, corresponding respectively to the two subunits of HIV-1 reverse transcriptase, in HIV samples. PA1-4948 immunogen corresponds to the HIV-1 Reverse Transcriptase which was over-expressed in E. coli as a recombinant protein. Recombinant HIV-1 Reverse Transcriptase protein (Cat. # RP-4912 or RP-4913) can be purchased for control experiments in Western blot assays.

HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1(AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 Reverse Transcriptase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.09% sodium azide
Gene Alias HIV RT
Host Species Rabbit
Immunogen Full size functional recombinant HIV 1 expressed and purified from E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.