Learn More
Invitrogen™ HIV1 Virions Polyclonal Antibody, FITC

Description
The PA1-73090 antibody reacts with HIV-1 Virions. PA1-73090 has been successfully used in Immunofluorescence applications. The PA1-73090 antibody was raised against HIV-1 isolate.
Specifications
Specifications
| Antigen | HIV1 Virions |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | HIV-1; HIV-1 Virions |
| Host Species | Goat |
| Immunogen | Human confirmed HIV-1 isolate. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.