Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV2 Nef Monoclonal Antibody (4B5)
HIV2 Nef Monoclonal Antibody (4B5)
Supplier: Thermo Scientific MA517015
Description

Specifications
HIV2 Nef | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Rat | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
4B5 | |
Unconjugated | |
HIV-2; HIV-2 nef; HIV2 nef protein; Nefprotein | |
TrpE-Fusion protein produced in X63Ag8.653 cells. | |
50 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction