Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIVEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546725 Supplier Invitrogen™ Supplier No. PA546725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FSYERSGYDL EESDGPDEDD NENEDDDEDS QAESVLSATP SVTASPQHLP Sequence homology: Cow: 100%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.

This gene encodes a transcription factor belonging to the ZAS family, members of which are large proteins that contain a ZAS domain - a modular protein structure consisting of a pair of C2H2 zinc fingers with an acidic-rich region and a serine/threonine-rich sequence. These proteins bind specifically to the DNA sequence motif, GGGACTTTCC, found in the enhancer elements of several viral promoters, including human immunodeficiency virus (HIV), and to related sequences found in the enhancer elements of a number of cellular promoters. This protein binds to this sequence motif, suggesting a role in the transcriptional regulation of both viral and cellular genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIVEP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HIVEP1
Gene Accession No. P15822
Gene Alias alpha A-CRYBP1; alpha A-crystallin-binding protein 1; alpha A-crystallin-binding protein I; alphaA-CRYBP1; cirhin interaction protein; CIRIP; Cryabp1; CRYBP1; crystallin, alpha binding protein 1; GAAP; Gate keeper of apoptosis-activating protein; HIV type I enhancer binding protein 1; HIVEP zinc finger 1; HIVEP1; HIV-EP1; human immunodeficiency virus type I enhancer binding protein 1; human immunodeficiency virus type I enhancer-binding protein 1; major histocompatibility complex binding protein 1; Major histocompatibility complex-binding protein 1; MBP-1; positive regulatory domain II binding factor 1; Positive regulatory domain II-binding factor 1; PRDII-BF1; Schnurri-1; si:dkey-184j23.2; si:dkey-31k9.2; transcription factor alphaA-CRYBP1; ZAS1; zinc finger protein 40; ZNF40; ZNF40A
Gene Symbols HIVEP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HIVEP1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3096
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.