Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIVEP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140120
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: CFSGVHTDPM DVLPRALLTR MTVLSTAQSD YNRKTLSPGK ARQRAARDEN DTIPSVDTSR SPCHQMSVDY PESEEILRSS MAGKAV.
This gene encodes a transcription factor belonging to the ZAS family, members of which are large proteins that contain a ZAS domain - a modular protein structure consisting of a pair of C2H2 zinc fingers with an acidic-rich region and a serine/threonine-rich sequence. These proteins bind specifically to the DNA sequence motif, GGGACTTTCC, found in the enhancer elements of several viral promoters, including human immunodeficiency virus (HIV), and to related sequences found in the enhancer elements of a number of cellular promoters. This protein binds to this sequence motif, suggesting a role in the transcriptional regulation of both viral and cellular genes.
Specifications
HIVEP1 | |
Polyclonal | |
Unconjugated | |
HIVEP1 | |
alpha A-CRYBP1; alpha A-crystallin-binding protein 1; alpha A-crystallin-binding protein I; alphaA-CRYBP1; cirhin interaction protein; CIRIP; Cryabp1; CRYBP1; crystallin, alpha binding protein 1; GAAP; Gate keeper of apoptosis-activating protein; HIV type I enhancer binding protein 1; HIVEP zinc finger 1; HIVEP1; HIV-EP1; human immunodeficiency virus type I enhancer binding protein 1; human immunodeficiency virus type I enhancer-binding protein 1; major histocompatibility complex binding protein 1; Major histocompatibility complex-binding protein 1; MBP-1; positive regulatory domain II binding factor 1; Positive regulatory domain II-binding factor 1; PRDII-BF1; Schnurri-1; si:dkey-184j23.2; si:dkey-31k9.2; transcription factor alphaA-CRYBP1; ZAS1; zinc finger protein 40; ZNF40; ZNF40A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3096 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P15822 | |
HIVEP1 | |
Recombinant protein corresponding to Human HIVEP1. Recombinant protein control fragment (Product #RP-109133). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction