Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIVEP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100756 Supplier Invitrogen™ Supplier No. PA5100756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MIBP1.

HIVEP2 is a member of a family of closely related, large, zinc finger-containing transcription factors. It regulates transcription by binding to regulatory regions of various cellular and viral genes that maybe involved in growth, development and metastasis.HIVEP2 contains the ZAS domain comprised of two widely separated regions of zinc finger motifs, a stretch of highly acidic amino acids and a serine/threonine-rich sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIVEP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HIVEP2
Gene Accession No. P31629, Q00900, Q3UHF7
Gene Alias Agie-bp1; Angiotensinogen gene-inducible enhancer-binding protein 1; c-myc intron binding protein 1; DNA-binding protein AGIE-BP1; Hivep2; HIV-EP2; human immunodeficiency virus type 1 enhancer-binding protein 2; human immunodeficiency virus type I enhancer binding protein 2; Human immunodeficiency virus type I enhancer-binding protein 2; Human immunodeficiency virus type I enhancer-binding protein 2 homolog; MBP-2; MHC binding protein-2; MHC-binding protein 2; MIBP1; MIBP-1; MRD43; Myc intron-binding protein 1; Schnurri-2; SHN2; Shn-2; transcription factor HIVEP2; ZAS2; ZNF40B
Gene Symbols HIVEP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human(unreviewed) HIVEP2(Accession P31629), corresponding to amino acid residues Q682-M711.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15273, 29721, 3097
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.