Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HJURP Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147288JF549
Description
HJURP Polyclonal antibody specifically detects HJURP in Human samples. It is validated for Western BlotSpecifications
HJURP | |
Polyclonal | |
Western Blot | |
FAKTS14-3-3-associated AKT substrate, fetal liver expressing gene 1, Fetal liver-expressing gene 1 protein, FLEG1, hFLEG1, Holliday junction recognition protein, Up-regulated in lung cancer 9, URLC9DKFZp762E1312 | |
The immunogen for this product maps to a region between residue 700 and 748 of human Holliday Junction Recognition Protein using the numbering given in entry NP_060880.3 (GeneID 55355). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55355 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction