Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HJURP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585663 Supplier Invitrogen™ Supplier No. PA585663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Positive Control: MDA-MB-231. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Holliday junction recognition protein (HJURP) is a centromeric protein that plays a central role in the incorporation and maintenance of histone H3-like variant CENPA at centromeres. It acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. HJURP prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. It directly binds to Holliday junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HJURP
Applications Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene HJURP
Gene Accession No. Q8NCD3
Gene Alias 14-3-3-associated AKT substrate; 6430706D22Rik; A730008H23Rik; C330011F01Rik; FAKTS; fetal liver expressing gene 1; fetal liver expressing gene 1 protein homolog; Fetal liver-expressing gene 1 protein; FLEG1; Gm975; hFLEG1; Hjurp; Holliday junction recognition protein; mFleg1; RGD1305846; Up-regulated in lung cancer 9; URLC9
Gene Symbols HJURP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human HJURP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.