Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580688 Supplier Invitrogen™ Supplier No. PA580688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human HK1/Hexokinase 1.

This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HK1
Gene Accession No. P19367
Gene Alias BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s
Gene Symbols HK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human HK1/Hexokinase 1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.