Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582278 Supplier Invitrogen™ Supplier No. PA582278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSGMYLGELV RLILVKMAKE GLLFEGRITP ELLTRGKFNT SDVSAIEKNK EGLHNAKEIL TRLGVEPSDD DCVSVQHVCT IVSFRSANLV AATLGAILNR LRDNKGTPRL RTTVGVDGSL YKTHPQYSRR FHKTLR Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 93%.

This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HK1
Gene Accession No. P05708, P17710, P19367
Gene Alias BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s
Gene Symbols HK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HK1. Recombinant protein control fragment (Product #RP-89762).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15275, 25058, 3098
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.