Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HK2 Monoclonal Antibody (3D3)
GREENER_CHOICE

Catalog No. PIMA515679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515679 Supplier Invitrogen™ Supplier No. MA515679
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15679 targets HK2 in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human samples. The MA5-15679 immunogen is purified recombinant fragment of human HK2 expressed in E. Coli. MA5-15679 detects HK2 which has a predicted molecular weight of approximately 102kDa.

In vertebrates there are four major glucose-phosphorylating isoenzymes, designated hexokinase I, II, III, and IV. Hexokinase activity is involved in the first step in several metabolic pathways including phosphorylation of glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Hexokinase is an allosteric enzyme inhibited by its product GLC-6-P. Hexokinase 2 is the predominant hexokinase isozyme expressed in insulin-responsive tissues such as skeletal muscle. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HK2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3D3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Hk2
Gene Accession No. P52789
Gene Alias AI642394; DKFZp686M1669; Hexokinase; hexokinase 2; hexokinase II; hexokinase type II; Hexokinase-2; hexokinase-2 muscle; hexokinase-2, muscle; Hexokinase-B; HK II; Hk2; HKII; HXK2; muscle form hexokinase; OTTHUMP00000202738
Gene Symbols Hk2
Host Species Mouse
Immunogen Purified recombinant fragment of human HK2 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.