Learn More
Invitrogen™ HK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121197
Description
Positive test controls include: HepG2, HL-60, THP-1, NCI-H460, Mouse liver. Immunogen sequence: MDSIGSSGLR QGEETLSCSE EGLPGPSDSS ELVQECLQQF KVTRAQLQQI QASLLGSMEQ ALRGQASPAP AVRMLPTYVG STPHGTEQGD FVVLELGATG ASLRVLWVTL TGIEGHRVEP RSQEFVIPQE VMLGAGQQLF DFAAHCLSEF LDAQPVNKQG LQLGFSFSFP CHQTGLDRST LISWTKGFRC SGVEGQDVVQ LLRDAIRRQG AYNIDVVAVV NDTVGTMMGC EPGVRPCEVG LVVDTGTNAC YMEEARHVAV LDEDRGRVCV.
Hexokinases phosphorylate glucose to produce glucose 6 phosphate, thus committing glucose to the glycolytic pathway. In vertebrates there are four major glucose phosphorylating isoenzymes, designated hexokinase I, II, III and IV (glucokinase). Similar to hexokinases I and II, the allosteric enzyme hexokinase III is inhibited by its product glucose 6 phosphate. The catalytic activity of hexokinase III is associated with the C terminus while regulatory function is associated with the N terminus.
Specifications
HK3 | |
Polyclonal | |
Unconjugated | |
HK3 | |
ATP:D-hexose 6-phosphotransferase; hexokinase 3; hexokinase 3 (white cell); hexokinase iii; hexokinase type III; Hexokinase3; Hexokinase-3; Hexokinase-C; HK III; HK3; HKIII; HK-III; HXK3; RNU73859 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
212032, 25060, 3101 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.83 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27926, P52790, Q3TRM8 | |
HK3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human HK3 (NP_002106.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.