Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HKDC1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP243539

Catalog No. NBP243539


Only null left
Add to Cart

Description

Description

Specifically detects HKDC1 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence

HKDC1 Polyclonal specifically detects HKDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HKDC1
Polyclonal
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
EC 2.7.1, EC 2.7.1.1, FLJ22761, FLJ37767, hexokinase domain containing 1, Hexokinase domain-containing protein 1, MGC125688, putative hexokinase HKDC1
Rabbit
103 kDa
0.1 mL
Primary
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
HKDC1
Recombinant protein encompassing a sequence within the center region of human HKDC1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
80201
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only